2,4-Difluoroaniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
D101400-10G D101400-25G
SIALD101400-10GEA 47.9 EUR
SIALD101400-10G SIALD101400-25G
2,4-Difluoroaniline, Sigma-Aldrich®
2,4-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 170
Melting Pt: –7 °C
Density: 1.268 g/cm³ (20 °C)
Flash Pt: 62
Storage Temperature: Ambient
MDL Number: MFCD00007648
CAS Number: 367-25-9
EINECS: 206-687-5
UN: 2941
ADR: 6.1 III
Merck Index: 13,03171