2,3,4,5,6-Pentafluoroaniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
103713-25G 103713-5G
SIAL103713-25GEA 99.1 EUR
SIAL103713-25G SIAL103713-5G
2,3,4,5,6-Pentafluoroaniline, Sigma-Aldrich®
2,3,4,5,6-Pentafluoroaniline
Formula: C₆H₂F₅N
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: 36…37 °C
Density: 1.744 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007643
CAS Number: 771-60-8
EINECS: 212-234-2