4-Fluoro-3-nitroaniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
155861-100G 155861-25G
SIAL155861-100GEA 121 EUR
SIAL155861-100G SIAL155861-25G
4-Fluoro-3-nitroaniline, Sigma-Aldrich®
4-Fluoro-3-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 96…98 °C
Density: 0.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007833
CAS Number: 364-76-1
EINECS: 206-665-5