3-(Difluoromethoxy)aniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
566446-1G 566446-5G
SIAL566446-1GEA 37.2 EUR
SIAL566446-1G SIAL566446-5G
3-(Difluoromethoxy)aniline, Sigma-Aldrich®
3-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
MDL Number: MFCD00236221
CAS Number: 22236-08-4