4-(Difluoromethoxy)aniline, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

470139-1G 470139-5G
SIAL470139-1GEA 61.5 EUR
SIAL470139-1G SIAL470139-5G
4-(Difluoromethoxy)aniline, Sigma-Aldrich®
4-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 90 °C (1013 hPa)
Density: 1.286 g/cm³ (20 °C)
MDL Number: MFCD00085005
CAS Number: 22236-10-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR