4-(Trifluoromethoxy)aniline
Supplier: Thermo Scientific
TL00398.1
TL00398.10
MAYBTL00398.1EA
0
EUR
MAYBTL00398.1
MAYBTL00398.10
4-(Trifluoromethoxy)aniline
4-(Trifluoromethoxy)aniline
Formula:
C₇H₆F₃NO Boiling Pt: 73 °C (1013 hPa) Density: 1.31 g/cm³ (20 °C) |
MDL Number:
MFCD00041314 CAS Number: 461-82-5 EINECS: 207-317-5 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...