Sanger's reagent, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene
M-Clarity™ quality level = MQ200
D1529-100ML
D1529-10ML
D1529-25ML
SIALD1529-100MLEA
453
EUR
SIALD1529-100ML
SIALD1529-10ML
SIALD1529-25ML
Sanger's reagent, Sigma-Aldrich®
Sanger's reagent
Formula:
C₆H₃FN₂O₄ MW: 186.1 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 24…26 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007056 CAS Number: 70-34-8 EINECS: 200-734-3 UN: 2811 ADR: 6.1,III Merck Index: 13,04199 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...