4-(Trifluoromethoxy)phenol 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: α,α,α-Trifluoro-4-hydroxyanisole
306040010 306040050
ACRO306040010EA 25.6 EUR
ACRO306040010 ACRO306040050
4-(Trifluoromethoxy)phenol 98%
4-(Trifluoromethoxy)phenol
Formula: C₇H₅F₃O₂
MW: 178.11 g/mol
Boiling Pt: 80 °C (15 mmHg)
Melting Pt: 17…18 °C
Density: 1.37 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00040988
CAS Number: 828-27-3
EINECS: 212-583-0
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4490 (20°C, 589 nm)
GC ≥97.5 %