4-(Trifluoromethoxy)phenol
Supplier: Thermo Scientific
Synonyms:
α,α,α-Trifluoro-4-hydroxyanisole
RDR03368.1
RDR03368.10
RDR03368.50
MAYBRDR03368.1EA
0
EUR
MAYBRDR03368.1
MAYBRDR03368.10
MAYBRDR03368.50
4-(Trifluoromethoxy)phenol
4-(Trifluoromethoxy)phenol
Formula:
C₇H₅F₃O₂ MW: 178.11 g/mol Boiling Pt: 180 °C (1013 hPa) Melting Pt: 17…18 °C Density: 1.37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00040988 CAS Number: 828-27-3 EINECS: 212-583-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...