4-(Trifluoromethoxy)phenol, Sigma-Aldrich®
Supplier: Merck
Synonyms: α,α,α-Trifluoro-4-hydroxyanisole
M-Clarity™ quality level = MQ200
339865-1G 339865-5G
SIAL339865-1GEA 21.1 EUR
SIAL339865-1G SIAL339865-5G
4-(Trifluoromethoxy)phenol, Sigma-Aldrich®
4-(Trifluoromethoxy)phenol
Formula: C₇H₅F₃O₂
MW: 178.11 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 17…18 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040988
CAS Number: 828-27-3
EINECS: 212-583-0
UN: 2810
ADR: 6.1,III