3,4-Difluorophenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
290408-25G 290408-5G
SIAL290408-25GEA 127 EUR
SIAL290408-25G SIAL290408-5G
3,4-Difluorophenol, Sigma-Aldrich®
3,4-Difluorophenol
Formula: C₆H₄F₂O
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 32…36 °C
MDL Number: MFCD00010315
CAS Number: 2713-33-9
UN: 2921
ADR: 8,II