2,5-Difluorophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
290394-1G 290394-5G
SIAL290394-1GEA 30.5 EUR
SIAL290394-1G SIAL290394-5G
2,5-Difluorophenol, Sigma-Aldrich®
2,5-Difluorophenol
Formula: C₆H₄F₂O
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: 42 °C
MDL Number: MFCD00042501
CAS Number: 2713-31-7
UN: 2921
ADR: 8,II