5-Fluoro-2-nitrophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Fluoro-2-hydroxynitrobenzene
M-Clarity™ quality level = MQ200
233242-100G 233242-25G
SIAL233242-100GEA 133 EUR
SIAL233242-100G SIAL233242-25G
5-Fluoro-2-nitrophenol, Sigma-Aldrich®
5-Fluoro-2-nitrophenol
Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 36…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00007107
CAS Number: 446-36-6
EINECS: 207-168-6