3,4-Difluorothiophenol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 3,4-Difluorobenzene-1-thiol, 3,4-Difluorobenzenethiol, 1,2-Difluoro-4-mercaptobenzene, 3,4-Difluorothiophenol

M-Clarity™ quality level = MQ100

549223-25G 549223-5G
SIAL549223-25GEA 510 EUR
SIAL549223-25G SIAL549223-5G
3,4-Difluorothiophenol, Sigma-Aldrich®
3,4-Difluorothiophenol
Formula: C₆H₄F₂S
MW: 146.16 g/mol
MDL Number: MFCD03094279
CAS Number: 60811-24-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR