2,4-Difluorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,4-Difluorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

548081-25G 548081-5G
SIAL548081-25GEA 53.2 EUR
SIAL548081-25G SIAL548081-5G
2,4-Difluorobenzenesulphonyl chloride, Sigma-Aldrich®
2,4-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 84 °C (1013 hPa)
Density: 1.581 g/cm³ (20 °C)
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR