2,6-Difluorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,6-Difluorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

544264-1G 544264-5G
SIAL544264-1GEA 33.5 EUR
SIAL544264-1G SIAL544264-5G
2,6-Difluorobenzenesulphonyl chloride, Sigma-Aldrich®
2,6-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 127.5…129.5 °C (1013 hPa)
Density: 1.568 g/cm³ (20 °C)
MDL Number: MFCD00085004
CAS Number: 60230-36-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR