3,4-Difluorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3,4-Difluorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

555916-1G 555916-5G
SIAL555916-1GEA 26.1 EUR
SIAL555916-1G SIAL555916-5G
3,4-Difluorobenzenesulphonyl chloride, Sigma-Aldrich®
3,4-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 105 °C (15 mmHg)
Density: 1.586 g/cm³ (20 °C)
MDL Number: MFCD00153120
CAS Number: 145758-05-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR