1,4-Diiodobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
193526-10G 193526-50G
SIAL193526-10GEA 66.8 EUR
SIAL193526-10G SIAL193526-50G
1,4-Diiodobenzene, Sigma-Aldrich®
1,4-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 126…127 °C
Density: 2.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2