Methyl-4-iodobenzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Iodobenzoic acid methyl ester, Methyl 4-iodobenzoate
M-Clarity™ quality level = MQ100
679100-25G 679100-5G
SIAL679100-25GEA 96.4 EUR
SIAL679100-25G SIAL679100-5G
Methyl-4-iodobenzoate, Sigma-Aldrich®
Methyl 4-iodobenzoate
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 276
Melting Pt: 112
Density: 2.02 g/cm³ (10 °C)
MDL Number: MFCD00016353
CAS Number: 619-44-3
EINECS: 210-597-1