2,3,5-Triiodobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: TIBA
120979-100G 120979-10G
SIAL120979-100GEA 357 EUR
SIAL120979-100G SIAL120979-10G
2,3,5-Triiodobenzoic acid, Sigma-Aldrich®
2,3,5-Triiodobenzoic acid
Formula: C₇H₃I₃O₂
MW: 499.81 g/mol
Boiling Pt: 416 °C (1013 hPa)
Melting Pt: 225.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00002420
CAS Number: 88-82-4
EINECS: 201-859-6