2,3,5-Triiodobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: TIBA
M-Clarity™ quality level = MQ200
T5910-10G T5910-5G
SIALT5910-10GEA 236 EUR
SIALT5910-10G SIALT5910-5G
2,3,5-Triiodobenzoic acid, Sigma-Aldrich®
2,3,5-Triiodobenzoic acid
Formula: C₇H₃I₃O₂
MW: 499.81 g/mol
Boiling Pt: 416 °C (1013 hPa)
Melting Pt: 225.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00002420
CAS Number: 88-82-4
EINECS: 201-859-6