4-Iodobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: p-Iodobenzoic acid
M-Clarity™ quality level = MQ200
206547-25G 206547-5G
SIAL206547-25GEA 83.7 EUR
SIAL206547-25G SIAL206547-5G
4-Iodobenzoic acid, Sigma-Aldrich®
4-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 319
Density: 2.184 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002533
CAS Number: 619-58-9
EINECS: 210-603-2