2-Iodoaniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
I7004-25G I7004-5G
SIALI7004-25GEA 123 EUR
SIALI7004-25G SIALI7004-5G
2-Iodoaniline, Sigma-Aldrich®
2-Iodoaniline
Formula: C₆H₆IN
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0