4-Iodoaniline, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

129364-100G 129364-25G
SIAL129364-100GEA 69.4 EUR
SIAL129364-100G SIAL129364-25G
4-Iodoaniline, Sigma-Aldrich®
4-Iodoaniline
Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00007848
CAS Number: 540-37-4
EINECS: 208-743-4
Merck Index: 13,05049

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR