1-Iodo-4-nitrobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
I9805-100G I9805-25G
SIALI9805-100GEA 453 EUR
SIALI9805-100G SIALI9805-25G
1-Iodo-4-nitrobenzene, Sigma-Aldrich®
1-Iodo-4-nitrobenzene
Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 289
Melting Pt: 168
Density: 2.273 g/cm³ (20 °C)
MDL Number: MFCD00007299
CAS Number: 636-98-6
EINECS: 211-272-7