4-Iodoanisole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
I7608-100G I7608-25G
SIALI7608-100GEA 122 EUR
SIALI7608-100G SIALI7608-25G
4-Iodoanisole, Sigma-Aldrich®
4-Iodoanisole
Formula: C₇H₇IO
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 48…49 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001056
CAS Number: 696-62-8
EINECS: 211-798-7