1,3-Dimethoxy-4-iodobenzene ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: 1-Iodo-2,4-dimethoxybenzene, 2,4-Dimethoxyiodobenzene
L19420.03 L19420.06
L19420.03EA 19.8 EUR
L19420.03 L19420.06
1,3-Dimethoxy-4-iodobenzene ≥97%
1,3-Dimethoxy-4-iodobenzene
Purity: 97% CAS No.: 20469-63-0 Molecular Formula: C8H9IO2 Formula Weight: 264.06 Flash Point: 110°(230°F) Refractive Index: 1.6100 Melting Point: 37-41° Boiling Point: 135°/14mm Beilstein Reference: 1945135 MDL No.: MFCD00059268
Formula: C₈H₉IO₂
MW: 264.06 g/mol
Boiling Pt: 135 °C (14 mmHg)
Melting Pt: 37…41 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059268
CAS Number: 20469-63-0