2-Iodophenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
281409-25G 281409-5G
SIAL281409-25GEA 85.7 EUR
SIAL281409-25G SIAL281409-5G
2-Iodophenol, Sigma-Aldrich®
2-Iodophenol
Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 186…187 °C (213 hPa)
Melting Pt: 39…42 °C
Density: 1.8757 g/cm³ (80 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013963
CAS Number: 533-58-4
EINECS: 208-569-9
UN: 2811
ADR: 6.1 III
Merck Index: 13,05057