4-Iodophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
I10201-10G I10201-25G
SIALI10201-10GEA 62 EUR
SIALI10201-10G SIALI10201-25G
4-Iodophenol, Sigma-Aldrich®
4-Iodophenol
Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 287
Melting Pt: 92…93 °C
Density: 2.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002327
CAS Number: 540-38-5
EINECS: 208-745-5
UN: 3261
ADR: 8 III
Merck Index: 13,05058