4-Iodobenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
p-Iodobenzenesulphonyl Chloride, Pipsyl chloride, 4-Iodobenzene-1-sulphonyl chloride
24416.0005
BV24416.0025
B24416.06
B24416.14
B24416.22
ABCR24416.0005EA
0
EUR
ABCR24416.0005
ABCRBV24416.0025
B24416.06
B24416.14
B24416.22
4-Iodobenzenesulphonyl chloride ≥97%
4-Iodobenzenesulphonyl chloride
Purity: 97%
CAS No.: 98-61-3
Molecular Formula: C6H4ClIO2S
Formula Weight: 302.52
Melting Point: 79-84°
Beilstein Reference: 2691661
Merck Reference: 14,7486
UN No.: 3261
MDL No.: MFCD00007441
Formula:
C₆H₄ClIO₂S Melting Pt: 79…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007441 CAS Number: 98-61-3 EINECS: 202-686-9 UN: 3261 ADR: 8,II Merck Index: 14,7486 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...