N,N-Diethylaniline, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

185868-1L 185868-250ML 185868-50ML
SIAL185868-1LEA 64.8 EUR
SIAL185868-1L SIAL185868-250ML SIAL185868-50ML
N,N-Diethylaniline, Sigma-Aldrich®
N,N-Diethylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 215.5 °C (1013 hPa)
Melting Pt: –38.8 °C
Density: 0.9348 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00009042
CAS Number: 91-66-7
EINECS: 202-088-8
UN: 2432
ADR: 6.1,III
Merck Index: 13,03140

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR