N,N-Dimethylaniline, Sigma-Aldrich®
Supplier: Merck
Danger
MQ200
515124-100ML 515124-1L 515124-2L 515124-4L
SIAL515124-100MLEA 25.4 EUR
SIAL515124-100ML SIAL515124-1L SIAL515124-2L SIAL515124-4L
N,N-Dimethylaniline, Sigma-Aldrich®
N,N-Dimethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 194
Melting Pt: 2.5
Density: 0.9559 g/cm³ (20 °C)
Flash Pt: 62
Storage Temperature: Ambient
MDL Number: MFCD00008304
CAS Number: 121-69-7
EINECS: 204-493-5
UN: 2253
ADR: 6.1 II
Merck Index: 13,03261