4-Amino-N,N-dimethylaniline, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
N,N-Dimethylbenzene-p-diamine, N,N-Dimethyl-1,4-phenylenediamine, N,N-Dimethylbenzene-1,4-diamine, N,N-Dimethyl-p-phenylenediamine
M-Clarity™ quality level = MQ200
193992-100G
193992-500G
193992-5G
SIAL193992-100GEA
144
EUR
SIAL193992-100G
SIAL193992-500G
SIAL193992-5G
4-Amino-N,N-dimethylaniline, Sigma-Aldrich®
4-Amino-N,N-dimethylaniline
Formula:
C₈H₁₂N₂ MW: 136.2 g/mol Boiling Pt: 262 °C (1013 hPa) Melting Pt: 35 °C Density: 1.09 g/cm³ (20 °C) Flash Pt: 130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007860 CAS Number: 99-98-9 EINECS: 202-807-5 UN: 2811 ADR: 6.1,II Merck Index: 13,33280 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...