3-Nitrophenyl isocyanate

Supplier: Thermo Scientific

Synonyms: 1-Isocyanato-3-nitrobenzene, 3-Nitrophenylisocyanate

SB01754.1 SB01754.10 SB01754.50
MAYBSB01754.1EA 0 EUR
MAYBSB01754.1 MAYBSB01754.10 MAYBSB01754.50
3-Nitrophenyl isocyanate
3-Nitrophenyl isocyanate
Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
Boiling Pt: 120…125 °C (0.7 mmHg)
Melting Pt: 52
Storage Temperature: Refrigerator
MDL Number: MFCD00007220
CAS Number: 3320-87-4
EINECS: 222-025-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR