4-Methoxyphenyl isocyanate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Isocyanato-4-methoxybenzene
M-Clarity™ quality level = MQ100
238600-25G 238600-5G
SIAL238600-25GEA 231 EUR
SIAL238600-25G SIAL238600-5G
4-Methoxyphenyl isocyanate, Sigma-Aldrich®
4-Methoxyphenyl isocyanate
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 106 °C (1013 hPa)
Density: 1.1559 g/cm³ (20 °C)
MDL Number: MFCD00002026
CAS Number: 5416-93-3
EINECS: 226-513-1
UN: 2206
ADR: 6.1,III