Phenyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
139742-100G 139742-500G
SIAL139742-100GEA 73.1 EUR
SIAL139742-100G SIAL139742-500G
Phenyl isothiocyanate, Sigma-Aldrich®
Phenyl isothiocyanate
Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 221
Melting Pt: –21 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Refrigerator
MDL Number: MFCD00004798
CAS Number: 103-72-0
EINECS: 203-138-1
UN: 2927
ADR: 6.1 II
Merck Index: 12,07450