3-Methoxyphenyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3-Methoxyphenylisothiocyanate
A15090.14 A15090.06 A15090.22
A15090.14EA 140 EUR
A15090.14 A15090.06 A15090.22
3-Methoxyphenyl isothiocyanate ≥98%
3-Methoxyphenyl isothiocyanate
Purity: 98% CAS No.: 3125-64-2 Molecular Formula: C8H7NOS Formula Weight: 165.22 Flash Point: 110°(230°F) Refractive Index: 1.6390 Odour: Pungent Boiling Point: 133-134°/10mm Beilstein Reference: 775987 UN No.: 2922 MDL No.: MFCD00040335
Formula: C₈H₇NOS
MW: 165.22 g/mol
Boiling Pt: 133…134 °C (10 mmHg)
Density: 1.179
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040335
CAS Number: 3125-64-2
ADR: 8 III