3-Methoxyphenyl isothiocyanate

Supplier: Thermo Scientific

Synonyms: 3-Methoxyphenylisothiocyanate

TL00330.1 TL00330EA TL00330.50
MAYBTL00330.1EA 0 EUR
MAYBTL00330.1 MAYBTL00330EA MAYBTL00330.50
3-Methoxyphenyl isothiocyanate
3-Methoxyphenyl isothiocyanate
Formula: C₈H₇NOS
MW: 165.22 g/mol
Boiling Pt: 84 °C (1 mmHg)
Density: 1,1788 g/cm³ (20 °C)
MDL Number: MFCD00040335
CAS Number: 3125-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR