4-(Methylthio)phenyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L09270.03
L09270.06
L09270.03EA
36.8
EUR
L09270.03
L09270.06
4-(Methylthio)phenyl isothiocyanate ≥97%
4-(Methylthio)phenyl isothiocyanate
Purity: 97%
CAS No.: 15863-41-9
Molecular Formula: C8H7NS2
Formula Weight: 181.28
Flash Point:
Formula:
C₈H₇NS₂ Boiling Pt: 127…128 °C (1 mmHg) Melting Pt: 48…51 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041368 CAS Number: 15863-41-9 UN: 2923 ADR: 8,III |