Aniline, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Aminobenzene, Phenylamine
M-Clarity™ quality level = MQ200
132934-1L 132934-2.5L
SIAL132934-1LEA 108 EUR
SIAL132934-1L SIAL132934-2.5L
Aniline, Sigma-Aldrich®
Aniline
Formula: C₆H₅NH₂
MW: 93.13 g/mol
Boiling Pt: 184
Melting Pt: –6.2 °C
Density: 1.019 g/cm³ (20 °C)
Flash Pt: 76
Storage Temperature: Ambient
MDL Number: MFCD00007629
CAS Number: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1 II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM