3-Nitroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: m-Nitroaniline
M-Clarity™ quality level = MQ100
N9829-100G N9829-500G N9829-5G
SIALN9829-100GEA 53.9 EUR
SIALN9829-100G SIALN9829-500G SIALN9829-5G
3-Nitroaniline, Sigma-Aldrich®
3-Nitroaniline
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 109…112 °C
Density: 0.9 g/cm³ (25 °C)
Flash Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD00007782
CAS Number: 99-09-2
EINECS: 202-729-1
UN: 1661
ADR: 6.1,II
Merck Index: 12,06679