4-Nitroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: p-Nitroaniline, pNA
M-Clarity™ quality level = MQ200
185310-100G 185310-500G 185310-5G
SIAL185310-100GEA 44.2 EUR
SIAL185310-100G SIAL185310-500G SIAL185310-5G
4-Nitroaniline, Sigma-Aldrich®
4-Nitroaniline
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 148 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00007858
CAS Number: 100-01-6
EINECS: 202-810-1
UN: 1661
ADR: 6.1,II
Merck Index: 12,06681