2-Amino-5-nitrophenol 95%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Hydroxy-4-nitroaniline
400902500 400900010 400900250 400900500
ACRO400902500EA 202 EUR
ACRO400902500 ACRO400900010 ACRO400900250 ACRO400900500
2-Amino-5-nitrophenol 95%
2-Amino-5-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 198…202 °C (dec.)
Density: 1,54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007692
CAS Number: 121-88-0
EINECS: 204-503-8
Order Now
Specification Test Results
Appearance Rust brown to brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
GC ≥96.0 %