2-Amino-4-nitrophenol
Supplier: Thermo Scientific
Warning
Synonyms: 2-Hydroxy-5-nitroaniline
SB01488.1 SB01488.10 SB01488EE
MAYBSB01488.1EA 0 EUR
MAYBSB01488.1 MAYBSB01488.10 MAYBSB01488EE
2-Amino-4-nitrophenol
2-Amino-4-nitrophenol
Formula: C₆H₆N₂O₃
Melting Pt: 176…177 °C
Density: 1.66 g/cm³ (20 °C)
MDL Number: MFCD00007695
CAS Number: 99-57-0
EINECS: 202-767-9