4-Amino-3-nitrophenol 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Hydroxy-2-nitroaniline
335220050 335220250 335221000
ACRO335220050EA 27.3 EUR
ACRO335220050 ACRO335220250 ACRO335221000
4-Amino-3-nitrophenol 98%
4-Amino-3-nitrophenol
Formula: C₆H₆N₂O₃
Melting Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00066310
CAS Number: 610-81-1
EINECS: 210-236-8
Order Now
Specification Test Results
Appearance Red to bordeaux Crystalline powder
Infrared spectrum Conforms
Melting point ≥150 °C
HPLC ≥97.5 %