4-Amino-3-nitrophenol 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Hydroxy-2-nitroaniline
L09487.06 L09487.14
L09487.06EA 36.9 EUR
L09487.06 L09487.14
4-Amino-3-nitrophenol 98%
4-Amino-3-nitrophenol
Purity: 98% CAS No.: 610-81-1 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Odour: Characteristic Melting Point: 148-153° Beilstein Reference: 2210196 MDL No.: MFCD00066310
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 148…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00066310
CAS Number: 610-81-1
EINECS: 210-236-8