(2-Aminoethoxy)benzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: ω-Aminophenetole, 2-Phenoxyethylamine, 2-Aminoethyl phenyl ether, β-Aminophenetole

448400-1G 448400-5G
SIAL448400-1GEA 57.2 EUR
SIAL448400-1G SIAL448400-5G
(2-Aminoethoxy)benzene, Sigma-Aldrich®
(2-Aminoethoxy)benzene
Formula: C₆H₅OCH₂CH₂NH₂
MW: 137.18 g/mol
Boiling Pt: 228…229 °C (1013 hPa)
Density: 1,048 g/cm³ (20 °C)
MDL Number: MFCD00052975
CAS Number: 1758-46-9
EINECS: 217-153-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR