o-Anisidine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Anisidine, 2-Aminoanisole, 2-Methoxyaniline
M-Clarity™ quality level = MQ200
A88182-5G A88182-100G A88182-500G
SIALA88182-5GEA 34.4 EUR
SIALA88182-5G SIALA88182-100G SIALA88182-500G
o-Anisidine, Sigma-Aldrich®
o-Anisidine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 225
Melting Pt: 5
Density: 1.09 g/cm³ (20 °C)
Flash Pt: 118
Storage Temperature: Ambient
MDL Number: MFCD00007688
CAS Number: 90-04-0
EINECS: 201-963-1
UN: 2431
ADR: 6.1 III
Merck Index: 13,00670