3,5-Dimethoxyaniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
D130001-25G D130001-5G
SIALD130001-25GEA 128 EUR
SIALD130001-25G SIALD130001-5G
3,5-Dimethoxyaniline, Sigma-Aldrich®
3,5-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8
EINECS: 233-616-5
UN: 2811
ADR: 6.1,III