2-Aminophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Amino-1-hydroxy benzene, 2-Hydroxy aniline, 2-Hydroxyaniline, o-Aminophenol
M-Clarity™ quality level = MQ200
A71301-100G A71301-500G A71301-5G
SIALA71301-100GEA 35.6 EUR
SIALA71301-100G SIALA71301-500G SIALA71301-5G
2-Aminophenol, Sigma-Aldrich®
2-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 235 °C (1013 hPa)
Density: 1.328 g/cm³ (20 °C)
Flash Pt: >175 °C
MDL Number: MFCD00007690
CAS Number: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 13,00460